Anti-SIRPA Rabbit Polyclonal Antibody

Supplier: Sino Biological
50956-T16-50 50956-T16-100 50956-T16-200
103589-382EA 142.96 USD
103589-382 103595-858 103595-860
Anti-SIRPA Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse SIRP alpha / CD172a (Catalog#50956-M08H; BAA20376.1; Met1-Asn373). SIRP alpha / CD172a specific IgG was purified by Mouse SIRP alpha / CD172a affinity chromatography.

Type: Primary
Antigen: SIRPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR