Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SIRPA Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 012]

Supplier: Sino Biological

50956-R012-F-100 50956-R012-F-25
103632-348EA 150.77 USD InStock
103632-348 103629-544
Anti-SIRPA Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 012]
Antibodies
Anti-SIRP alpha Antibody (FITC

Type: Primary
Antigen: SIRPA
Clonality: Monoclonal
Clone: Clone: 012
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty