Anti-SIRPA Rabbit Monoclonal Antibody [clone:012]

Supplier: Sino Biological
50956-R012-50 50956-R012-100
103659-710EA 271.4 USD
103659-710 103595-856
Anti-SIRPA Rabbit Monoclonal Antibody [clone:012]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SIRPA/SIRP alpha/CD172a (rM SIRPA/SIRP alpha/CD172a; Catalog#50956-M08H; BAA20376.1; Met1-Asn373).

Type: Primary
Antigen: SIRPA
Clonality: Monoclonal
Clone: 012
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR