Anti-SIRPA Rabbit Monoclonal Antibody [clone: 009]

Supplier: Sino Biological

50956-R009-50 50956-R009-100
103595-902EA 128.15 USD
103595-902 103659-708
Anti-SIRPA Rabbit Monoclonal Antibody [clone: 009]
Antibodies
Anti-SIRP alpha Antibody

Type: Primary
Antigen: SIRPA
Clonality: Monoclonal
Clone: Clone: 009
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR