Anti-VNN1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50952-RP02-100 50952-RP02-200 50952-RP02-50
103659-694EA 270.87 USD
103659-694 103659-696 103595-892
Anti-VNN1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse VNN1 / Vanin-1 (rM VNN1 / Vanin-1; Catalog#50952-M08H; Q9Z0K8; Met1-Ser487). VNN1 / Vanin-1 specific IgG was purified by Mouse VNN1 / Vanin-1 affinity chromatography.

Type: Primary
Antigen: VNN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR