Anti-VNN1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50952-RP01-100 50952-RP01-400 50952-RP01-200
103659-690EA 106.62 USD
103659-690 103659-692 103659-688
Anti-VNN1 Rabbit Polyclonal Antibody
Antibodies
Anti-VNN1 Antibody

Type: Primary
Antigen: VNN1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR