Anti-NAALADL1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50937-RP01-200 50937-RP01-100 50937-RP01-400
103659-546EA 160.46 USD
103659-546 103659-548 103659-550
Anti-NAALADL1 Rabbit Polyclonal Antibody
Antibodies
Anti-NAALADL1 Antibody

Type: Primary
Antigen: NAALADL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR