Anti-CNTN1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50933-T16-100 50933-T16-50 50933-T16-200
103589-372EA 142.96 USD
103589-372 103595-868 103595-870
Anti-CNTN1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Contactin 1 / CNTN1 (rM Contactin 1 / CNTN1; Catalog#50933-M08H; NP_031753.1; Met1-Leu1000). Contactin 1 / CNTN1 specific IgG was purified by Mouse Contactin 1 / CNTN1 affinity chromatography.

Type: Primary
Antigen: Contactin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR