Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CHI3L1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50929-RP01-100 50929-RP01-200 50929-RP01-400
103659-678EA 106.62 USD InStock
103659-678 103659-680 103659-682
Anti-CHI3L1 Rabbit Polyclonal Antibody
Antibodies
Anti-CHI3L1 Antibody

Type: Primary
Antigen: CHI3L1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty