Anti-TACSTD2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50922-RP02-200 50922-RP02-50 50922-RP02-100
103659-660EA 270.87 USD
103659-660 103659-662 103594-656
Anti-TACSTD2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse TROP-2 / TACSTD2 (rM TROP-2 / TACSTD2; Catalog#50922-M08H; Q8BGV3; Met1-Gln270). TROP-2 / TACSTD2 specific IgG was purified by Mouse TROP-2 / TACSTD2 affinity chromatography.

Type: Primary
Antigen: TROP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR