Anti-TACSTD2 Rabbit Monoclonal Antibody [clone:064]

Supplier: Sino Biological
50922-R064-100 50922-R064-50
103659-656EA 301.66 USD
103659-656 103595-852
Anti-TACSTD2 Rabbit Monoclonal Antibody [clone:064]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TROP-2 / TACSTD2 (Catalog#50922-M08H; Q8BGV3; Met1-Gln270).

Type: Primary
Antigen: TROP2
Clonality: Monoclonal
Clone: 064
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR