Anti-KLK7 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50921-T24-200 50921-T24-50 50921-T24-100
103595-848EA 352.03 USD
103595-848 103595-846 103589-368
Anti-KLK7 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse KLK7/Kallikrein 7(PRSS6) (rM KLK7/Kallikrein 7(PRSS6); Catalog#50921-M08H; Q91VE3; Met1-Arg249). KLK7/Kallikrein 7(PRSS6) specific IgG was purified by Mouse KLK7/Kallikrein 7(PRSS6) affinity chromatography.

Type: Primary
Antigen: KLK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR