Anti-CST6 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50916-RP02-100 50916-RP02-50 50916-RP02-200
103595-836EA 142.96 USD
103595-836 103659-644 103659-646
Anti-CST6 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Cystatin-E / CST6 / Cystatin-M / Cystatin-6 (rM Cystatin-E / CST6 / Cystatin-M / Cystatin-6; Catalog#50916-M08H; NP_082899.1; Met1-Ala152). Cystatin-E / CST6 / Cystatin-M / Cystatin-6 specific IgG was purified by Mouse Cystatin-E / CST6 / Cystatin-M / Cystatin-6 affinity chromatography.

Type: Primary
Antigen: CST6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR