Anti-BAMBI Rabbit Polyclonal Antibody

Supplier: Sino Biological
50895-T16-100 50895-T16-50 50895-T16-200
103595-824EA 352.03 USD
103595-824 103595-822 103589-358
Anti-BAMBI Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse BAMBI / NMA (Catalog#50895-M08H; Q9D0L6; Met1-Ala152). BAMBI / NMA specific IgG was purified by Mouse BAMBI / NMA affinity chromatography.

Type: Primary
Antigen: BAMBI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR