Anti-CSK Rabbit Polyclonal Antibody

Supplier: Sino Biological

50893-T48-200 50893-T48-100 50893-T48-50
103595-816EA 214.31 USD
103595-816 103595-814 103589-354
Anti-CSK Rabbit Polyclonal Antibody
Antibodies
Anti-CSK Antibody

Type: Primary
Antigen: CSK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR