Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EPCR Rabbit Polyclonal Antibody

Supplier: Sino Biological

50874-T16-200 50874-T16-50 50874-T16-100
103595-800EA 214.31 USD InStock
103595-800 103589-346 103595-798
Anti-EPCR Rabbit Polyclonal Antibody
Antibodies
Anti-EPCR Antibody

Type: Primary
Antigen: EPCR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty