Anti-EPCR Rabbit Polyclonal Antibody

Supplier: Sino Biological

50874-RP01-400 50874-RP01-100 50874-RP01-200
103659-628EA 214.31 USD
103659-628 103659-624 103659-626
Anti-EPCR Rabbit Polyclonal Antibody
Antibodies
Anti-EPCR Antibody

Type: Primary
Antigen: EPCR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR