Anti-EPCR Rabbit Monoclonal Antibody (APC (Allophycocyanin)) [clone: 016]

Supplier: Sino Biological

50874-R016-A-25 50874-R016-A-100
103630-172EA 86.15 USD
103630-172 103632-336
Anti-EPCR Rabbit Monoclonal Antibody (APC (Allophycocyanin)) [clone: 016]
Antibodies
Anti-EPCR Antibody (APC)

Type: Primary
Antigen: EPCR
Clonality: Monoclonal
Clone: Clone: 016
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR