Anti-FASLG Rabbit Monoclonal Antibody [clone: 001]

Supplier: Sino Biological

50854-R001-100 50854-R001-50
103659-616EA 214.31 USD
103659-616 103595-786
Anti-FASLG Rabbit Monoclonal Antibody [clone: 001]
Antibodies
Anti-Fas Ligand Antibody

Type: Primary
Antigen: FASLG
Clonality: Monoclonal
Clone: Clone: 001
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR