Anti-IL10RB Rabbit Polyclonal Antibody

Supplier: Sino Biological

50846-RP02-50 50846-RP02-200 50846-RP02-100
103595-780EA 106.62 USD
103595-780 103659-614 103659-612
Anti-IL10RB Rabbit Polyclonal Antibody
Antibodies
Anti-IL10RB Antibody

Type: Primary
Antigen: IL10RB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR