Anti-KLRC1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50834-T48-100 50834-T48-50 50834-T48-200
103595-768EA 352.03 USD
103595-768 103595-766 103589-336
Anti-KLRC1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse NKG2A / NKG2 / CD159A / KLRC1 (rM NKG2A / NKG2 / CD159A / KLRC1; Catalog#50834-M07H; AAD24969.1; Ala94-Ile244). NKG2A / NKG2 / CD159A / KLRC1 specific IgG was purified by Mouse NKG2A / NKG2 / CD159A / KLRC1 affinity chromatography.

Type: Primary
Antigen: NKG2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR