Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NKG2A Rabbit Polyclonal Antibody

Supplier: Sino Biological

50834-RP01-200 50834-RP01-100 50834-RP01-400
103660-752EA 160.46 USD InStock
103660-752 103660-750 103659-586
Anti-NKG2A Rabbit Polyclonal Antibody
Antibodies
Anti-NKG2A Antibody

Type: Primary
Antigen: NKG2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty