Anti-THOP1 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50828-RP02-100 50828-RP02-50 50828-RP02-200
103660-748EA 352.03 USD
103660-748 103660-746 103595-764
Anti-THOP1 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse THOP1 (rM THOP1; Catalog#50828-M07B; NP_073144.3; Lys 2-Cys687). THOP1 specific IgG was purified by Mouse THOP1 affinity chromatography.

Type: Primary
Antigen: THOP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR