Anti-THOP1 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50828-RP01-100 50828-RP01-400 50828-RP01-200
103659-580EA 106.62 USD
103659-580 103659-584 103659-582
Anti-THOP1 Rabbit Polyclonal Antibody
Antibodies
Anti-Thimet Oligopeptidase Ant

Type: Primary
Antigen: THOP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR