Anti-CDC37 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50826-T60-100 50826-T60-200 50826-T60-50
103595-754EA 160.46 USD
103595-754 103595-756 103589-332
Anti-CDC37 Rabbit Polyclonal Antibody
Antibodies
Anti-CDC37 Antibody

Type: Primary
Antigen: CDC37
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR