Anti-SIRPB Rabbit Polyclonal Antibody

Supplier: Sino Biological

50816-T16-200 50816-T16-100 50816-T16-50
103595-740EA 214.31 USD
103595-740 103595-738 103589-326
Anti-SIRPB Rabbit Polyclonal Antibody
Antibodies
Anti-SIRP-beta Antibody

Type: Primary
Antigen: SIRPB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR