Anti-SLC3A2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50813-RP02-100 50813-RP02-200 50813-RP02-50
103595-734EA 142.96 USD
103595-734 103659-038 103659-040
Anti-SLC3A2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse SLC3A2 (rM SLC3A2; Catalog#50813-M07H; NP_001154885.1; Ala 139-Ala 565). SLC3A2 specific IgG was purified by Mouse SLC3A2 affinity chromatography.

Type: Primary
Antigen: CD98
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR