Anti-SLC3A2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50813-RP01-100 50813-RP01-400 50813-RP01-200
103659-032EA 229.88 USD
103659-032 103659-034 103659-036
Anti-SLC3A2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse CD98 / SLC3A2 (Catalog#50813-M07H; NP_001154885.1; Ala139-Ala565). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: CD98
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR