Anti-IL1R1 Rabbit Monoclonal Antibody [clone:191] APC (Allophycocyanin)

Supplier: Sino Biological
50807-R191-A-100 50807-R191-A-25
103632-538EA 70.7 USD
103632-538 103630-166
Anti-IL1R1 Rabbit Monoclonal Antibody [clone:191] APC (Allophycocyanin)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-1R1 / CD121a (rM IL-1R1 / CD121a; Catalog#50807-M08H; P13504; Met1-Lys338) and conjugated with APC under optimum conditions, the unreacted APC was removed.

Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: Clone: 191
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR