Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL1R1 Rabbit Monoclonal Antibody [clone: 191]

Supplier: Sino Biological

50807-R191-50 50807-R191-100
103595-724EA 128.15 USD InStock
103595-724 103659-020
Anti-IL1R1 Rabbit Monoclonal Antibody [clone: 191]
Antibodies
Anti-IL1R1 Antibody

Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: Clone: 191
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty