Anti-IL1R1 Rabbit Monoclonal Antibody [clone: 075]

Supplier: Sino Biological

50807-R075-50 50807-R075-100
103595-722EA 128.15 USD
103595-722 103659-018
Anti-IL1R1 Rabbit Monoclonal Antibody [clone: 075]
Antibodies
Anti-IL1R1 Antibody

Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: Clone: 075
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR