Anti-IL1R1 Rabbit Monoclonal Antibody [clone:002]

Supplier: Sino Biological
50807-R002-1 50807-R002-100 50807-R002-50
103659-016EA 271.4 USD
103659-016 103589-322 103595-720
Anti-IL1R1 Rabbit Monoclonal Antibody [clone:002]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-1R1 / CD121a (rM IL-1R1 / CD121a; Catalog#50807-M08H; P13504; Met1-Lys338).

Type: Primary
Antigen: IL1R1
Clonality: Monoclonal
Clone: 002
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR