Anti-Klrb1a Rabbit Monoclonal Antibody [clone: 008]

Supplier: Sino Biological

50806-R008-50 50806-R008-100
103595-714EA 128.15 USD
103595-714 103659-008
Anti-Klrb1a Rabbit Monoclonal Antibody [clone: 008]
Antibodies
Anti-Klrb1a Antibody

Type: Primary
Antigen: Klrb1a
Clonality: Monoclonal
Clone: Clone: 008
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR