Anti-PDCD1LG2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50804-RP01-200 50804-RP01-400 50804-RP01-100
103659-002EA 270.2 USD
103659-002 103659-004 103659-006
Anti-PDCD1LG2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse B7-DC / PD-L2 / CD273 (Catalog#50804-M08H; Q9WUL5; Met1-Arg219). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: PD-L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR