Anti-TCPTP Rabbit Polyclonal Antibody

Supplier: Sino Biological
50800-T52-200 50800-T52-50 50800-T52-100
103595-708EA 210.04 USD
103595-708 103595-706 103589-316
Anti-TCPTP Rabbit Polyclonal Antibody
Antibodies
Anti-TCPTP Antibody

Type: Primary
Antigen: TCPTP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR