Anti-PTPN2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50800-RP01-200 50800-RP01-100 50800-RP01-400
103659-000EA 562.97 USD
103659-000 103659-532 103658-998
Anti-PTPN2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse PTPN2 / TC-PTP (Catalog#50800-M07B; Q06180-1; Ser2-Asn314). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: TCPTP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR