Anti-PTPN2 Rabbit Monoclonal Antibody [clone:001]

Supplier: Sino Biological
50800-R001-50 50800-R001-100
103659-530EA 271.4 USD
103659-530 103595-704
Anti-PTPN2 Rabbit Monoclonal Antibody [clone:001]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PTPN2 / TC-PTP (rM PTPN2 / TC-PTP; Catalog#50800-M07B; Q06180-1; Ser2-Asn314).

Type: Primary
Antigen: TCPTP
Clonality: Monoclonal
Clone: 001
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR