Anti-DDC Rabbit Polyclonal Antibody

Supplier: Sino Biological
50799-T56-50 50799-T56-100 50799-T56-200
103595-702EA 352.03 USD
103595-702 103595-700 103589-314
Anti-DDC Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse DOPA Decarboxylase / DDC (Catalog#50799-M08B; O88533; Met1-Glu480). DOPA Decarboxylase / DDC specific IgG was purified by Mouse DOPA Decarboxylase / DDC affinity chromatography.

Type: Primary
Antigen: DDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR