Anti-DDC Rabbit Polyclonal Antibody

Supplier: Sino Biological

50799-RP01-200 50799-RP01-100 50799-RP01-400
103659-524EA 160.46 USD
103659-524 103659-526 103659-528
Anti-DDC Rabbit Polyclonal Antibody
Antibodies
Anti-DDC Antibody

Type: Primary
Antigen: DDC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR