Anti-MCAM Rabbit Polyclonal Antibody

Supplier: Sino Biological

50794-RP02-50 50794-RP02-200 50794-RP02-100
103595-698EA 106.62 USD
103595-698 103659-522 103659-520
Anti-MCAM Rabbit Polyclonal Antibody
Antibodies
Anti-CD146 / MCAM Antibody

Type: Primary
Antigen: MCAM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR