Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MCAM Rabbit Polyclonal Antibody

Supplier: Sino Biological

50794-RP01-200 50794-RP01-100 50794-RP01-400
103658-994EA 160.46 USD InStock
103658-994 103658-996 103659-518
Anti-MCAM Rabbit Polyclonal Antibody
Antibodies
Anti-CD146 / MCAM Antibody

Type: Primary
Antigen: MCAM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty