Anti-LAMP2 Rabbit Polyclonal Antibody

Supplier: Sino Biological

50791-T48-100 50791-T48-50 50791-T48-200
103595-692EA 160.46 USD
103595-692 103589-312 103595-694
Anti-LAMP2 Rabbit Polyclonal Antibody
Antibodies
Anti-LAMP2 Antibody

Type: Primary
Antigen: LAMP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR