Anti-LAMP2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50791-RP02-100 50791-RP02-200 50791-RP02-50
103595-690EA 99 USD
103595-690 103660-742 103660-740
Anti-LAMP2 Rabbit Polyclonal Antibody
Antibodies
Anti-LAMP2 Antibody

Type: Primary
Antigen: LAMP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR