Anti-LAMP2 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50791-RP01-100 50791-RP01-200 50791-RP01-400
103658-992EA 351.49 USD
103658-992 103658-990 103660-738
Anti-LAMP2 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse LAMP2 / CD107b (Catalog#50791-M08H; P17047-1; Leu26-Asn379). Total IgG was purified by Protein A affinity chromatography.

Type: Primary
Antigen: LAMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR