Anti-LAMP2 Rabbit Monoclonal Antibody [clone:141]

Supplier: Sino Biological
50791-R141-50 50791-R141-100
103595-688EA 150.18 USD
103595-688 103658-988
Anti-LAMP2 Rabbit Monoclonal Antibody [clone:141]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LAMP2/CD107b (rM LAMP2/CD107b; Catalog#50791-M08H; P17047-1; Leu26-Asn379).

Type: Primary
Antigen: LAMP2
Clonality: Monoclonal
Clone: 141
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR