Anti-CPA2 Rabbit Monoclonal Antibody [clone:179]

Supplier: Sino Biological
50778-R179-100 50778-R179-50
103595-680EA 150.18 USD
103595-680 103659-514
Anti-CPA2 Rabbit Monoclonal Antibody [clone:179]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carboxypeptidase A2 / CPA2 (rM Carboxypeptidase A2 / CPA2; Catalog#50778-M08H; Q504N0; Met1-Tyr417).

Type: Primary
Antigen: CPA2
Clonality: Monoclonal
Clone: 179
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR