Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CPA2 Rabbit Monoclonal Antibody [clone: 133]

Supplier: Sino Biological

50778-R133-50 50778-R133-100
103595-678EA 128.15 USD InStock
103595-678 103659-512
Anti-CPA2 Rabbit Monoclonal Antibody [clone: 133]
Antibodies
Anti-Carboxypeptidase A2 Antib

Type: Primary
Antigen: CPA2
Clonality: Monoclonal
Clone: Clone: 133
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty