Anti-SELPLG Rabbit Monoclonal Antibody [clone:001] FITC (Fluorescein Isothiocyanate)

Supplier: Sino Biological
50770-R001-F-100 50770-R001-F-25
103632-514EA 70.7 USD
103632-514 103630-158
Anti-SELPLG Rabbit Monoclonal Antibody [clone:001] FITC (Fluorescein Isothiocyanate)
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PSGL-1/CD162 (rM PSGL-1/CD162; Catalog#50770-MCCH; Q62170; Met1-Cys307) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.

Type: Primary
Antigen: PSGL-1
Clonality: Monoclonal
Clone: Clone: 001
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR