Anti-CA10 Rabbit Polyclonal Antibody

Supplier: Sino Biological
50768-RP02-100 50768-RP02-200 50768-RP02-50
103595-658EA 142.96 USD
103595-658 103658-956 103658-958
Anti-CA10 Rabbit Polyclonal Antibody
Antibodies
Produced in rabbits immunized with purified, recombinant Mouse Carbonic Anhydrase X / CA10 (rM Carbonic Anhydrase X / CA10; Catalog#50768-M08H; P61215; Met1-Asn300). Carbonic Anhydrase X / CA10 specific IgG was purified by Mouse Carbonic Anhydrase X / CA10 affinity chromatography.

Type: Primary
Antigen: CA10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR