Anti-CA10 Rabbit Monoclonal Antibody [clone:018]

Supplier: Sino Biological
50768-R018-50 50768-R018-100
103595-656EA 150.18 USD
103595-656 103658-948
Anti-CA10 Rabbit Monoclonal Antibody [clone:018]
Antibodies
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Carbonic Anhydrase X / CA10 (rM Carbonic Anhydrase X / CA10; Catalog#50768-M08H; P61215; Met1-Asn300).

Type: Primary
Antigen: CA10
Clonality: Monoclonal
Clone: 018
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR